1-Bromo-6-phenylhexane 97%

Supplier: Ambeed
Warning

Synonyms: (6-Bromohex-1-yl)benzene

A865741-1G A865741-250MG A865741-5G
76728-902EA 65.78 USD
76728-902 76728-904 76728-906
1-Bromo-6-phenylhexane 97%
1-Bromo-6-phenylhexane

Precaución: for Research Use Only

Formula: C₆H₅(CH₂)₆Br
MW: 241.18 g/mol
MDL Number: MFCD00236022
CAS Number: 27976-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR