N6-Benzyladenosine 98%

Supplier: Ambeed
Warning

A858213-1G A858213-250MG A858213-5G
76728-894EA 35 USD
76728-894 76728-896 76728-898
N6-Benzyladenosine 98%
N6-Benzyladenosine

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₅O₄
MDL Number: MFCD00005740
CAS Number: 4294-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR