Thioxanthen-9-one 95%

Supplier: Ambeed
Warning

Synonyms: Thioxanthone

A857980-100G A857980-10G A857980-1G A857980-25G A857980-500G A857980-5G
76728-882EA 128.37 USD
76728-882 76728-884 76728-886 76728-888 76728-890 76728-892
Thioxanthen-9-one 95%
Thioxanthen-9-one

Precaución: for Research Use Only

Formula: C₁₃H₈OS
MW: 212.2700 g/mol
MDL Number: MFCD00005066
CAS Number: 492-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR