Methyltributylphosphonium iodide 98%

Supplier: Ambeed
Warning

A854008-1G A854008-25G A854008-5G
76728-860EA 6 USD
76728-860 76728-862 76728-864
Methyltributylphosphonium iodide 98%
Methyltributylphosphonium iodide

Precaución: for Research Use Only

Formula: C₁₃H₃₀IP
MDL Number: MFCD00031626
CAS Number: 1702-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR