N-Acetyl-DL-methionine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ac-DL-Met-OH
A849460-100G
A849460-25G
A849460-500G
76728-848EA
10.61
USD
76728-848
76728-850
76728-852
N-Acetyl-DL-methionine 98%
N-Acetyl-DL-methionine
Precaución: for Research Use Only
Formula:
C₇H₁₃NO₃S MW: 191.2 g/mol |
MDL Number:
MFCD00008681 CAS Number: 1115-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...