Bis(8-quinolinolato)copper(II) 97%

Supplier: Ambeed
Warning

Synonyms: Bis(8-hydroxyquinolinato)copper(II), Oxine-Copper, Copper(II) Bis(8-hydroxyquinolinate)

A849325-1000G A849325-100G A849325-10G A849325-25G A849325-500G
76728-838EA 284.33 USD
76728-838 76728-840 76728-842 76728-844 76728-846
Bis(8-quinolinolato)copper(II) 97%
Bis(8-quinolinolato)copper(II)

Precaución: for Research Use Only

Formula: C₁₈H₁₂CuN₂O₂
MW: 351.85 g/mol
MDL Number: MFCD00067392
CAS Number: 10380-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR