1,2,3,4-Tetra-O-acetyl-β-D-xylopyranose 95%
Supplier: Ambeed
|
Warning
|
A846662-100G
A846662-1G
A846662-250MG
A846662-25G
A846662-5G
76728-826EA
185
USD
76728-826
76728-828
76728-830
76728-832
76728-834
1,2,3,4-Tetra-O-acetyl-β-D-xylopyranose 95%
1,2,3,4-Tetra-O-acetyl-beta-D-xylopyranose
Precaución: for Research Use Only
Formula:
C₁₃H₁₈O₉ |
MDL Number:
MFCD00069790 CAS Number: 4049-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...