N,N-Dimethyl-2-naphthylamine 98%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethylnaphthalen-2-amine, 2-Dimethylaminonaphthalene

A843793-1G A843793-250MG A843793-5G
76728-800EA 20 USD
76728-800 76728-802 76728-804
N,N-Dimethyl-2-naphthylamine 98%
N,N-Dimethyl-2-naphthylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₃N
MDL Number: MFCD00059550
CAS Number: 2436-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR