1,1,3,3-Tetrabutylurea 99%

Supplier: Ambeed
Warning

A842830-100G A842830-500G
76728-794EA 10.61 USD
76728-794 76728-796
1,1,3,3-Tetrabutylurea 99%
1,1,3,3-Tetrabutylurea

Precaución: for Research Use Only

Formula: C₁₇H₃₆N₂O
MW: 284.49 g/mol
MDL Number: MFCD00042901
CAS Number: 4559-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR