2-Piperidylacetic acid hydrochloride 97%

Supplier: Ambeed
Danger

A834927-1G A834927-250MG A834927-5G
76728-764EA 30 USD
76728-764 76728-766 76728-768
2-Piperidylacetic acid hydrochloride 97%
2-Piperidylacetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂·ClH
MDL Number: MFCD08061346
CAS Number: 19615-30-6
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR