N-(4-Fluorobenzylidene)aniline 98%

Supplier: Ambeed
Warning

A833248-250MG A833248-5G A833248-1G
76728-756EA 44 USD
76728-756 76728-758 77880-636
N-(4-Fluorobenzylidene)aniline 98%
N-(4-Fluorobenzylidene)aniline

Precaución: for Research Use Only

Formula: C₁₃H₁₀FN
MDL Number: MFCD00017971
CAS Number: 5676-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR