Methoxatin Disodium 98%

Supplier: Ambeed
Warning

Synonyms: Pyrroloquinoline quinone disodium salt

A831583-100MG A831583-10MG A831583-1G A831583-1MG A831583-25MG A831583-50MG A831583-5MG
76728-738EA 394.66 USD
76728-738 76728-740 76728-742 76728-744 76728-746 76728-748 76728-750
Methoxatin Disodium 98%
Methoxatin Disodium

Precaución: for Research Use Only

Formula: C₁₄H₄N₂Na₂O₈
MW: 374.16 g/mol
MDL Number: MFCD00151711
CAS Number: 122628-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR