Silver iodide 99+%

Supplier: Ambeed
Danger

A831269-25G A831269-5G
76728-728EA 35 USD
76728-728 76728-730
Silver iodide 99+%
Silver iodide

Precaución: for Research Use Only

Formula: AgI
MDL Number: MFCD00003412
CAS Number: 7783-96-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR