Nerolidol (cis and trans mixture) 97% (mixture of isomers)
Supplier: Ambeed
|
Danger
|
A825725-25G
A825725-1000G
A825725-100G
A825725-500G
76728-718EA
7
USD
76728-718
77670-174
77670-176
77670-178
Nerolidol (cis and trans mixture) 97% (mixture of isomers)
Nerolidol (cis and trans mixture)
Precaución: for Research Use Only
Formula:
C₁₅H₂₆O |
MDL Number:
MFCD00008911 CAS Number: 7212-44-4 UN: 3082 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...