Methyl-4-bromo-1-naphthoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromonaphthalene-1-carboxylate

A824457-1G A824457-5G
76728-714EA 26.53 USD
76728-714 76728-716
Methyl-4-bromo-1-naphthoate 97%
Methyl 4-bromo-1-naphthoate

Precaución: for Research Use Only

Formula: C₁₂H₉BrO₂
MW: 265.1027 g/mol
MDL Number: MFCD09475914
CAS Number: 35615-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR