n-Propyltriphenylphosphonium bromide 99%

Supplier: Ambeed
Warning

A822691-1000G A822691-100G A822691-25G A822691-500G
76728-706EA 131 USD
76728-706 76728-708 76728-710 76728-712
n-Propyltriphenylphosphonium bromide 99%
n-Propyltriphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₁H₂₂BrP
MDL Number: MFCD01090228
CAS Number: 15912-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR