N,N-Diethyl-m-toluidine 99%

Supplier: Ambeed
Warning

Synonyms: 3-Diethylaminotoluene, N,N-Diethyl-3-methylaniline, DMT

A822299-100G A822299-25G A822299-500G
76728-700EA 18 USD
76728-700 76728-702 76728-704
N,N-Diethyl-m-toluidine 99%
N,N-Diethyl-m-toluidine

Precaución: for Research Use Only

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Boiling Pt: 97
MDL Number: MFCD00035795
CAS Number: 91-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR