Dodecacarbonyltriruthenium 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Triruthenium dodecacarbonyl, Ruthenium carbonyl
A820757-1G
A820757-250MG
A820757-25G
A820757-5G
76728-686EA
53
USD
76728-686
76728-688
76728-690
76728-692
Dodecacarbonyltriruthenium 98%
Dodecacarbonyltriruthenium
Precaución: for Research Use Only
Formula:
C₁₂O₁₂Ru₃ |
MDL Number:
MFCD00011209 CAS Number: 15243-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...