N-(tert-Butoxycarbonyloxy)phthalimide 95+%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (1,3-dioxoisoindolin-2-yl) carbonate, Carbonic Acid tert-Butyl Phthalimido Ester, tert-Butyl Phthalimido Carbonate

A818248-1G A818248-5G
76728-682EA 38.2 USD
76728-682 76728-684
N-(tert-Butoxycarbonyloxy)phthalimide 95+%
N-(tert-Butoxycarbonyloxy)phthalimide

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₅
MW: 263.25 g/mol
MDL Number: MFCD00005889
CAS Number: 15263-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR