Mitotane 99%

Supplier: Ambeed
Danger

Synonyms: 1-Chloro-2-(2,2-dichloro-1-(4-chlorophenyl)ethyl)benzene, o,p'-DDD, 2,4'-TDE, 1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2-dichloroethane, o,p'-TDE, 2,4'-DDD, 1-Chloro-2-[2,2-dichloro-1-(4-chlorophenyl)ethyl]benzene

A817688-1MG A817688-25G A817688-5G A817688-1G
76728-678EA 5.31 USD
76728-678 76728-680 77603-080 77670-158
Mitotane 99%
Mitotane

Precaución: for Research Use Only

Formula: C₁₄H₁₀Cl₄
MW: 320.04 g/mol
MDL Number: MFCD00000850
CAS Number: 53-19-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR