Methyl 5-bromo-4-methylthiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

A814436-1G A814436-25G A814436-5G
76728-660EA 31 USD
76728-660 77603-058 77880-614
Methyl 5-bromo-4-methylthiophene-2-carboxylate 98%
Methyl 5-bromo-4-methylthiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇BrO₂S
MDL Number: MFCD04038959
CAS Number: 54796-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR