(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Aminomethylphenylboronic acid, pinacol ester
A813793-1G
A813793-250MG
A813793-5G
A813793-100MG
76728-654EA
66.84
USD
76728-654
76728-656
76728-658
77603-056
(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine 95%
(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine
Precaución: for Research Use Only
Formula:
C₁₃H₂₀BNO₂ MW: 233.11 g/mol |
MDL Number:
MFCD06213229 CAS Number: 138500-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...