Morpholine-4-carboxamidine hydrobromide 98%

Supplier: Ambeed
Warning

Synonyms: Morpholine-4-carboximidamide hydrobromide

A809849-5G A809849-25G
76728-652EA 28.65 USD
76728-652 77573-588
Morpholine-4-carboxamidine hydrobromide 98%
Morpholine-4-carboxamidine hydrobromide

Precaución: for Research Use Only

Formula: C₅H₁₂BrN₃O
MW: 210.075 g/mol
MDL Number: MFCD00051994
CAS Number: 157415-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR