N,N-Dimethyl-4-fluoro-2-nitroaniline 99%

Supplier: Ambeed
Warning

A809451-100G A809451-1G A809451-25G A809451-5G
76728-644EA 275 USD
76728-644 76728-646 76728-648 76728-650
N,N-Dimethyl-4-fluoro-2-nitroaniline 99%
N,N-Dimethyl-4-fluoro-2-nitroaniline

Precaución: for Research Use Only

Formula: C₈H₉FN₂O₂
MDL Number: MFCD04116334
CAS Number: 69261-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR