2-Phenyl-2-(2-pyridyl)acetamide 95%

Supplier: Ambeed
Warning

Synonyms: 2-Phenyl-2-(pyridin-2-yl)acetamide

A808871-100G A808871-10G A808871-25G A808871-500G
76728-636EA 41.38 USD
76728-636 76728-638 76728-640 76728-642
2-Phenyl-2-(2-pyridyl)acetamide 95%
2-Phenyl-2-(2-pyridyl)acetamide

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O
MW: 212.25 g/mol
MDL Number: MFCD00138126
CAS Number: 7251-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR