3-Benzylpiperidine 96%

Supplier: Ambeed
Danger

A808864-1G A808864-250MG A808864-100MG A808864-5G
76728-632EA 209 USD
76728-632 76728-634 77880-608 77880-609
3-Benzylpiperidine 96%
3-Benzylpiperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₇N
MDL Number: MFCD03426127
CAS Number: 13603-25-3
UN: 3267
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR