4-(Phenylsulfonamido)benzoic acid 95%

Supplier: Ambeed
Warning

A807557-1G A807557-250MG A807557-25G A807557-5G
76728-616EA 34 USD
76728-616 76728-618 76728-620 76728-622
4-(Phenylsulfonamido)benzoic acid 95%
4-(Phenylsulfonamido)benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₄S
MDL Number: MFCD00531525
CAS Number: 28547-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR