Pentafluorobenzoyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 2,3,4,5,6-Pentafluorobenzoyl chloride, Pentafluorobenzoylchloride, PFBC

A805157-1G A805157-25G A805157-5G
76728-612EA 11 USD
76728-612 76728-614 77552-480
Pentafluorobenzoyl chloride 95%
Pentafluorobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇ClF₅O
MW: 230.52 g/mol
Boiling Pt: 158
MDL Number: MFCD00000657
CAS Number: 2251-50-5
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR