Methyl-5-methylisoxazole-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-methylisoxazole-3-carboxylate

A805075-1G A805075-25G A805075-5G
76728-606EA 7.43 USD
76728-606 76728-608 76728-610
Methyl-5-methylisoxazole-3-carboxylate 98%
Methyl 5-methylisoxazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₃
MW: 141.13 g/mol
MDL Number: MFCD00015895
CAS Number: 19788-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR