Tetrapotassium diphosphate 99%

Supplier: Ambeed
Warning

Synonyms: Tetrapotassium diphosphate, Potassium diphosphate, Potassium pyrophosphate, tetra-Potassium pyrophosphate, Potassium pyrophospate

A803791-1000G A803791-500G A803791-100G
76728-602EA 8.49 USD
76728-602 76728-604 77670-134
Tetrapotassium diphosphate 99%
Tetrapotassium diphosphate

Precaución: for Research Use Only

Formula: K₄O₇P₂
MW: 330.34 g/mol
MDL Number: MFCD00011393
CAS Number: 7320-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR