Trisodium naphthalene-1,3,6-trisulfonate hydrate 65%

Supplier: Ambeed
Warning

A803718-100G A803718-25G A803718-500G
76728-596EA 27.59 USD
76728-596 76728-598 76728-600
Trisodium naphthalene-1,3,6-trisulfonate hydrate 65%
Trisodium naphthalene-1,3,6-trisulphonate hydrate

Precaución: for Research Use Only

Formula: C₁₀H₈O₉S₃·xH₂O
MDL Number: MFCD00003989
CAS Number: 5182-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR