Picolinic acid N-oxide 98%

Supplier: Ambeed
Warning

A803415-1G A803415-25G A803415-5G
76728-590EA 14.86 USD
76728-590 76728-592 76728-594
Picolinic acid N-oxide 98%
Picolinic acid N-oxide

Precaución: for Research Use Only

Formula: C₆H₅NO₃
MW: 139.1088 g/mol
MDL Number: MFCD00006198
CAS Number: 824-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR