Sodium bromodifluoroacetate 98%

Supplier: Ambeed
Warning

A802732-100G A802732-25G A802732-5G
76728-584EA 101 USD
76728-584 76728-586 76728-588
Sodium bromodifluoroacetate 98%
Sodium bromodifluoroacetate

Precaución: for Research Use Only

Formula: C₂HBrF₂O₂
MDL Number: MFCD04038355
CAS Number: 84349-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR