Phthalonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 1,2-Benzenedicarbonitrile, Benzene-1,2-dicarbonitrile, 1,2-Dicyanobenzene

A802090-25G A802090-100G
76728-582EA 7.43 USD
76728-582 77573-568
Phthalonitrile 95%
Phthalonitrile

Precaución: for Research Use Only

Formula: C₈H₄N₂
MW: 128.13 g/mol
MDL Number: MFCD00001771
CAS Number: 91-15-6
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR