n-Propyl cinnamate 97%

Supplier: Ambeed
Warning

Synonyms: Propyl 3-Phenylacrylate

A801753-10G A801753-25G A801753-5G
76728-576EA 11 USD
76728-576 76728-578 76728-580
n-Propyl cinnamate 97%
n-Propyl cinnamate

Precaución: for Research Use Only

Formula: C₆H₅CH=CHCO₂(CH₂)₂CH₃
MDL Number: MFCD00051556
CAS Number: 7778-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR