N,N'-Diphenylbenzidine 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Bis(phenylamino)biphenyl, N4,N4'-Diphenyl-[1,1'-biphenyl]-4,4'-diamine

A800662-10G A800662-1G A800662-25G A800662-5G
76728-554EA 18.04 USD
76728-554 76728-556 76728-558 76728-560
N,N'-Diphenylbenzidine 98%
N,N'-Diphenylbenzidine

Precaución: for Research Use Only

Formula: C₂₄H₂₀N₂
MW: 336.44 g/mol
MDL Number: MFCD00003016
CAS Number: 531-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR