1,3-Diethyl-1,3-diphenylurea 97%

Supplier: Ambeed
Warning

A799597-100G A799597-25G A799597-5G
76728-548EA 122 USD
76728-548 76728-550 76728-552
1,3-Diethyl-1,3-diphenylurea 97%
1,3-Diethyl-1,3-diphenylurea

Precaución: for Research Use Only

Formula: C₁₇H₂₀N₂O
MDL Number: MFCD00035794
CAS Number: 85-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR