Resazurin sodium salt 90+%

Supplier: Ambeed
Warning

Synonyms: 7-Sodiooxy-3H-phenoxazin-3-one 10-oxide, Sodium 7-oxido-3-oxo-3H-phenoxazine 10-oxide, Resazurin sodium, Resazurin

A798364-1G A798364-25G A798364-5G
76728-536EA 14 USD
76728-536 76728-538 76728-540
Resazurin sodium salt 90+%
Resazurin sodium salt

Precaución: for Research Use Only

Formula: C₁₂H₆NNaO₄
MW: 251.17 g/mol
MDL Number: MFCD00005036
CAS Number: 62758-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR