(2R,3R,4S,5S)-2-((Benzoyloxy)methyl)-5-methoxytetrahydrofuran-3,4-diyl dibenzoate 98%

Supplier: Ambeed
Warning

A798148-1G A798148-250MG
76728-532EA 47.75 USD
76728-532 76728-534
(2R,3R,4S,5S)-2-((Benzoyloxy)methyl)-5-methoxytetrahydrofuran-3,4-diyl dibenzoate 98%
(2R,3R,4S,5S)-2-((Benzoyloxy)methyl)-5-methoxytetrahydrofuran-3,4-diyl dibenzoate

Precaución: for Research Use Only

Formula: C₂₇H₂₄O₈
MW: 476.47 g/mol
MDL Number: MFCD00057268
CAS Number: 7473-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR