Methyl-5-methoxynicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-methoxypyridine-3-carboxylate, Methyl 5-methoxynicotinate

A797875-100MG A797875-1G A797875-250MG
76728-522EA 15 USD
76728-522 76728-524 76728-526
Methyl-5-methoxynicotinate 98%
Methyl-5-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MDL Number: MFCD09265109
CAS Number: 29681-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR