Propidium iodide 95%

Supplier: Ambeed
Warning

A795950-100MG A795950-250MG A795950-50MG A795950-25MG
76728-508EA 132 USD
76728-508 76728-510 76728-512 77573-546
Propidium iodide 95%
Propidium iodide

Precaución: for Research Use Only

Formula: C₂₇H₃₄I₂N₄
MDL Number: MFCD00011921
CAS Number: 25535-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR