S-Ethylisothiouronium bromide 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl carbamimidothioate hydrobromide, S-Ethylisothiourea Hydrobromide, S-EIT hydrobromide, 2-Ethyl-2-thiopseudourea hydrobromide

A793027-100G A793027-100MG A793027-1G A793027-250MG A793027-25G A793027-5G A793027-500G
76728-480EA 67 USD
76728-480 76728-482 76728-484 76728-486 76728-488 76728-490 77873-255
S-Ethylisothiouronium bromide 98%
S-Ethylisothiouronium bromide

Precaución: for Research Use Only

Formula: C₃H₉BrN₂S
MDL Number: MFCD00012585
CAS Number: 1071-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR