Methyl pyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

A791147-100MG A791147-250MG
76728-446EA 23.34 USD
76728-446 76728-448
Methyl pyrimidine-5-carboxylate 95%
Methyl pyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MW: 138.124 g/mol
MDL Number: MFCD06204205
CAS Number: 34253-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR