7,4'-Dihydroxy-6-methoxyisoflavone-7-D-glucoside 98%

Supplier: Ambeed
Warning

Synonyms: Glycitin, 3-(4-Hydroxyphenyl)-6-methoxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

A790821-10MG A790821-1MG A790821-25MG A790821-50MG A790821-5MG A790821-250MG
76728-436EA 25.47 USD
76728-436 76728-438 76728-440 76728-442 76728-444 77670-116
7,4'-Dihydroxy-6-methoxyisoflavone-7-D-glucoside 98%
7,4'-Dihydroxy-6-methoxyisoflavone-7-D-glucoside

Precaución: for Research Use Only

Formula: C₂₂H₂₂O₁₀
MW: 446.41 g/mol
MDL Number: MFCD00800711
CAS Number: 40246-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR