2-(4-(Bromomethyl)phenyl)-5,5-dimethyl-1,3,2-dioxaborinane 95+%

Supplier: Ambeed
Danger

A790672-1G A790672-250MG
76728-432EA 50 USD
76728-432 76728-434
2-(4-(Bromomethyl)phenyl)-5,5-dimethyl-1,3,2-dioxaborinane 95+%
2-(4-(Bromomethyl)phenyl)-5,5-dimethyl-1,3,2-dioxaborinane

Precaución: for Research Use Only

Formula: C₁₂H₁₆BBrO₂
MDL Number: MFCD03085921
CAS Number: 143805-78-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR