N-(2-Hydroxyethyl)methacrylamide 96% stabilized

Supplier: Ambeed
Warning

Synonyms: HEMAm, N-(2-Hydroxyethyl)-2-methylprop-2-enamide

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A789366-1G A789366-250MG A789366-25G
76728-426EA 45.62 USD
76728-426 76728-428 77552-436
N-(2-Hydroxyethyl)methacrylamide 96% stabilized
N-(2-Hydroxyethyl)methacrylamide

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.157 g/mol
MDL Number: MFCD00080529
CAS Number: 5238-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR