Methyl 4-amino-2-methoxy-5-nitrobenzoate 95%

Supplier: Ambeed
Warning

A788483-100MG A788483-1G A788483-250MG
76728-414EA 40.32 USD
76728-414 76728-416 76728-418
Methyl 4-amino-2-methoxy-5-nitrobenzoate 95%
Methyl 4-amino-2-methoxy-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₅
MW: 226.18 g/mol
MDL Number: MFCD09952401
CAS Number: 59338-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR