Benzyl ethyl malonate 97%

Supplier: Ambeed
Warning

A787062-1G A787062-250MG A787062-5G A787062-25G
76728-398EA 40.32 USD
76728-398 76728-400 76728-402 77670-108
Benzyl ethyl malonate 97%
Benzyl ethyl malonate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
MDL Number: MFCD00009194
CAS Number: 42998-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR