1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Diaza-18-crown-6, 1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane
A785615-100MG
A785615-1G
A785615-250MG
A785615-5G
A785615-25G
76728-378EA
24
USD
76728-378
76728-380
76728-382
76728-384
77670-104
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) 98%
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22)
Precaución: for Research Use Only
Formula:
C₁₂H₂₆N₂O₄ MW: 262.35 g/mol |
MDL Number:
MFCD00005112 CAS Number: 23978-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...