Methyl-4-methyl-1H-pyrrole-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-methyl-1H-pyrrole-2-carboxylate

A785542-100MG A785542-250MG A785542-5G A785542-1G
76728-372EA 48.81 USD
76728-372 76728-374 76728-376 77308-326
Methyl-4-methyl-1H-pyrrole-2-carboxylate 95%
Methyl-4-methyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.16 g/mol
MDL Number: MFCD00057179
CAS Number: 34402-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR